APO455826495 Apollo Scientific Ltd

4-Chloro-3-methoxyaniline

  • MDL number MFCD00672967
  • Cas Number : 13726-14-2
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Synonym 5-Amino-2-chloroanisole, 4-Chloro-m-anisidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-methoxyaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-methoxyaniline
pictograms GHS06;GHS08
hstatements H301+H311;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart