APO455826498 Apollo Scientific Ltd

3,5-Dibromo-4-methylbenzoic acid

  • MDL number MFCD00017544
  • Cas Number : 67973-32-4
  • Molecular Weight : 293.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dibromo-4-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dibromo-4-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart