APO455826500 Apollo Scientific Ltd

2-Nitro-N-(chloroacetyl)aniline

  • MDL number MFCD00024201
  • Cas Number : 10147-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-Chloro-2-nitroacetanilide, 2-Chloro-N-(nitrophenyl)acetamide
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26617

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26617
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart