APO455826501 Apollo Scientific Ltd

Methyl trans-3,4,5-trimethoxycinnamate

  • MDL number MFCD03614740
  • Cas Number : 20329-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Synonym Methyl trans-3-(3,4,5-trimethoxyphenyl)prop-2-enoate, Methyl (2E)-3-(3,4,5-trimethoxyphenyl)acrylate
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26620

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26620
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart