APO455826502 Apollo Scientific Ltd

1,3-Bis(3,4-dichlorophenyl)urea

  • MDL number MFCD00171464
  • Cas Number : 4300-43-0
  • Molecular Weight : 350.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 281-282
Synonym 3,3',4,4'-Tetrachlorocarbanilide
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/13-bis34-dichlorophenylurea

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-bis34-dichlorophenylurea
pictograms GHS07;GHS09
hstatements H319;H335;H400;H410
Availability details for
Quantity
  • Availability
  • Add to Cart