APO455826503 Apollo Scientific Ltd

2,6-Dichlorophenylthiourea

  • MDL number MFCD00041156
  • Cas Number : 6590-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159
Synonym 1,3-Dichloro-2-thioureidobenzene, 5-(Carbamothioylamino)-1,3-dichlorobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=33333

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=33333
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart