APO455826518 Apollo Scientific Ltd

5,6-Dichloro-1H-benzimidazole

  • MDL number MFCD00211325
  • Cas Number : 6478-73-5
  • Molecular Weight : 187.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-206
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/56-dichloro-1h-benzimidazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/56-dichloro-1h-benzimidazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart