APO455826523 Apollo Scientific Ltd

3-Hydroxy-2-methylbenzoic acid

  • MDL number MFCD00671541
  • Cas Number : 603-80-5
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 142-144
Iupac Name 3-hydroxy-2-methylbenzoic acid
Inchi Code 1S/C8H8O3/c1-5-6(8(10)11)3-2-4-7(5)9/h2-4,9H,1H3,(H,10,11)
Inchi Key RIERSGULWXEJKL-UHFFFAOYSA-N

Description

Synonym 3-Carboxy-2-methylphenol, 2-Carboxy-6-hydroxytoluene, 3-Hydroxy-o-toluic acid
Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 142-144
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-2-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-2-methylbenzoic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart