APO455826536 Apollo Scientific Ltd

5-Nitroindane

  • MDL number MFCD00003801
  • Cas Number : 7436-07-9
  • Molecular Weight : 163.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-nitroindane
Inchi Code 1S/C9H9NO2/c11-10(12)9-5-4-7-2-1-3-8(7)6-9/h4-6H,1-3H2
Inchi Key DLURUQQMVLOLCP-UHFFFAOYSA-N

Description

Synonym 2,3-Dihydro-5-nitro-1H-indene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-nitroindane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-nitroindane
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart