APO455826542 Apollo Scientific Ltd

2-Chloro-4,6-dimethylpyrimidine

  • MDL number MFCD00023199
  • Cas Number : 4472-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-38
Synonym 2-Chloro-4,6-dimethyl-1,3-diazine
Flash Point 110
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 210-212
Product Link https://store.apolloscientific.co.uk/product/2-chloro-46-dimethylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-46-dimethylpyrimidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart