APO455826543 Apollo Scientific Ltd

5-Cyanothiophene-2-boronic acid

  • MDL number MFCD02094029
  • Cas Number : 305832-67-1
  • Molecular Weight : 152.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-128
Synonym 5-Boronothiophene-2-carbonitrile
Purity 97
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-cyanothiophene-2-boronic-acid

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS https://store.apolloscientific.co.uk/product/5-cyanothiophene-2-boronic-acid
pictograms GHS06
hstatements H301+H311+H331;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart