APO455826559 Apollo Scientific Ltd

3,5-Diacetoxybenzoic acid

  • MDL number MFCD00017591
  • Cas Number : 35354-29-1
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 158-160
Iupac Name 3,5-bis(acetyloxy)benzoic acid
Inchi Code 1S/C11H10O6/c1-6(12)16-9-3-8(11(14)15)4-10(5-9)17-7(2)13/h3-5H,1-2H3,(H,14,15)
Inchi Key QBTDQJMLMVEUTQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 158-160
Product Link https://store.apolloscientific.co.uk/product/35-diacetoxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-diacetoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart