APO455826564 Apollo Scientific Ltd

4-Iodoquinoline

  • MDL number MFCD07700252
  • Cas Number : 16560-43-3
  • Molecular Weight : 255.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 94-96
Iupac Name 4-iodoquinoline
Inchi Code 1S/C9H6IN/c10-8-5-6-11-9-4-2-1-3-7(8)9/h1-6H
Inchi Key STERKHRCDZPNRE-UHFFFAOYSA-N

Description

Synonym 4-Iodo-1-azanaphthalene
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 94-96
Product Link https://store.apolloscientific.co.uk/product/4-iodoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart