APO455826576 Apollo Scientific Ltd

3-(3,5-Dimethoxyphenyl)propanoic acid

  • MDL number MFCD03425685
  • Cas Number : 717-94-2
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(3,5-dimethoxyphenyl)propanoic acid
Inchi Code 1S/C11H14O4/c1-14-9-5-8(3-4-11(12)13)6-10(7-9)15-2/h5-7H,3-4H2,1-2H3,(H,12,13)
Inchi Key LMBOJOXVLORKSQ-UHFFFAOYSA-N

Description

Synonym 3-(3,5-Dimethoxyphenyl)propionic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-35-dimethoxyphenylpropanoic-acid

Safety Information

signalword Warning
pcodes P280;P271;P264
MSDS https://store.apolloscientific.co.uk/product/3-35-dimethoxyphenylpropanoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart