APO455826578 Apollo Scientific Ltd

3-Methoxy-4-methylbenzaldehyde

  • MDL number MFCD04114050
  • Cas Number : 24973-22-6
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 40-42
Iupac Name 3-methoxy-4-methylbenzaldehyde
Inchi Code 1S/C9H10O2/c1-7-3-4-8(6-10)5-9(7)11-2/h3-6H,1-2H3
Inchi Key TVDHPUFLDYYBPO-UHFFFAOYSA-N

Description

Synonym 4-Formyl-2-methoxytoluene, 5-Formyl-2-methylanisole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 40-42
Product Link https://store.apolloscientific.co.uk/product/3-methoxy-4-methylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxy-4-methylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart