APO455826582 Apollo Scientific Ltd

5-Bromo-3-methylbenzo[b]thiophene

  • MDL number MFCD05664866
  • Cas Number : 1196-09-4
  • Molecular Weight : 227.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-39
Synonym 5-Bromo-3-methy-1-benzothiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 153-158/12mm
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-methylbenzobthiophene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-methylbenzobthiophene
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart