APO455826589 Apollo Scientific Ltd

4-(4-Methoxyphenoxy)aniline

  • MDL number MFCD00044105
  • Cas Number : 31465-36-8
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-79
Synonym 4-(4-Aminophenoxy)anisole, 4-Amino-4'-methoxydiphenyl ether
Purity 98
Density 1.294
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 245-250/33mm
Product Link https://store.apolloscientific.co.uk/product/4-4-methoxyphenoxyaniline

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-4-methoxyphenoxyaniline
pictograms GHS07;GHS08
hstatements H302+H312+H332;H334
Availability details for
Quantity
  • Availability
  • Add to Cart