APO455826593 Apollo Scientific Ltd

4-(3,4-Dichlorophenoxy)aniline

  • MDL number MFCD00459613
  • Cas Number : 67651-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68.7-70.3
Synonym 4-Amino-3',4'-dichlorodiphenyl ether
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27729

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27729
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart