APO455826596 Apollo Scientific Ltd

4-Chloro-4'-iododiphenyl ether

  • MDL number MFCD06796299
  • Cas Number : 854257-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-63
Synonym 1-Chloro-4-(4-iodophenoxy)benzene
Purity 99
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=27739

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=27739
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart