APO455826597 Apollo Scientific Ltd

4-Iodo-4'-methyldiphenyl ether

  • MDL number MFCD06796301
  • Cas Number : 1017793-91-7
  • Molecular Weight : 310.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Iodo-4-(4-methylphenoxy)benzene, 4-(4-Iodophenoxy)toluene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-4-methyldiphenyl-ether

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/4-iodo-4-methyldiphenyl-ether
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart