APO455826611 Apollo Scientific Ltd

7-Bromo-1H-indole, N-BOC protected

  • MDL number MFCD09475841
  • Cas Number : 868561-17-5
  • Molecular Weight : 296.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 7-bromo-1H-indole-1-carboxylate
Purity 98
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-1h-indole-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-1h-indole-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart