APO455826618 Apollo Scientific Ltd

[(2E)-3-(Dimethylamino)-1-methoxyprop-2-en-1-ylidene]malononitrile

  • MDL number MFCD00975440
  • Cas Number : 95689-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (E)-4,4-Dicyano-3-methoxy-N,N-dimethylbuta-1,3-dien-1-amine
Purity 98
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2e-3-dimethylamino-1-methoxyprop-2-en-1-ylidenemalononitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2e-3-dimethylamino-1-methoxyprop-2-en-1-ylidenemalononitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart