APO455826619 Apollo Scientific Ltd

4-[2-(Morpholin-4-yl)ethoxy]benzeneboronic acid, pinacol ester

  • MDL number MFCD04440857
  • Cas Number : 690636-28-3
  • Molecular Weight : 333.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-{2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]ethyl}morpholine, 2-{4-[2-(Morpholin-4-yl)ethoxy]phenyl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-2-morpholin-4-ylethoxybenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-2-morpholin-4-ylethoxybenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart