APO455826620 Apollo Scientific Ltd

Ethyl 7-nitro-1H-indole-2-carboxylate

  • MDL number MFCD00216475
  • Cas Number : 6960-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Synonym 2-(Ethoxycarbonyl)-7-nitro-1H-indole
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14683

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14683
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart