APO455826621 Apollo Scientific Ltd

tert-Butyl 4-bromo-2-nitrobenzoate

  • MDL number MFCD09475812
  • Cas Number : 890315-72-7
  • Molecular Weight : 302.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-4-bromo-2-nitrobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-4-bromo-2-nitrobenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart