APO455826632 Apollo Scientific Ltd

3-Bromo-2-chlorotoluene

  • MDL number MFCD07782165
  • Cas Number : 97329-43-6
  • Molecular Weight : 205.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 18
Synonym 1-Bromo-2-chloro-3-methylbenzene, 2-Chloro-3-methylbromobenzene
Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 125-135/ 50mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-chlorotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-chlorotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart