APO455826643 Apollo Scientific Ltd

2-Chloro-4-(pyrrolidin-1-yl)pyrimidine

  • MDL number MFCD09475891
  • Cas Number : 35691-20-4
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-122
Synonym 1-(2-Chloropyrimidin-4-yl)pyrrolidine
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-pyrrolidin-1-ylpyrimidine

Safety Information

signalword Warning
pcodes P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-pyrrolidin-1-ylpyrimidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart