APO455826649 Apollo Scientific Ltd

3-Bromo-N-methylbenzamide

  • MDL number MFCD01060879
  • Cas Number : 49834-22-2
  • Molecular Weight : 214.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-n-methylbenzamide

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-n-methylbenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart