APO455826653 Apollo Scientific Ltd

5-Bromo-4-(pent-1-yl)pyrimidine

  • MDL number MFCD09800934
  • Cas Number : 951884-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-4-(pent-1-yl)-1,3-diazine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40154

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40154
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart