APO455826658 Apollo Scientific Ltd

4-Bromo-2-methoxypyridine

  • MDL number MFCD08277268
  • Cas Number : 100367-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25-26
Storage temp. Store at 2-8
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methoxypyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methoxypyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart