APO455826659 Apollo Scientific Ltd

2-Amino-6-chloropyrazine

  • MDL number MFCD00055024
  • Cas Number : 33332-28-4
  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-152
Synonym 6-Chloropyrazin-2-amine
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-chloropyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-chloropyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart