APO455826665 Apollo Scientific Ltd

5-Bromo-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD08275714
  • Cas Number : 103878-58-6
  • Molecular Weight : 208.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-187
Synonym 5-Bromo-4-carboxy-1,3-thiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-13-thiazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-13-thiazole-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart