APO455826666 Apollo Scientific Ltd

3-Chloro-6-(methylamino)pyridazine

  • MDL number MFCD02178601
  • Cas Number : 14959-32-1
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-201
Synonym 6-Chloro-N-methylpyridazin-3-amine, 3-Chloro-6-(methylamino)-1,2-diazine
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-6-methylaminopyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-6-methylaminopyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart