APO455826668 Apollo Scientific Ltd

3-Chloro-5-[(morpholin-4-yl)carbonyl]benzeneboronic acid

  • MDL number MFCD09800860
  • Cas Number : 957120-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-146
Synonym (3-Borono-5-chlorophenyl)(morpholin-4-yl)methanone, 4-(3-Borono-5-chlorobenzoyl)morpholine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40478

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40478
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart