APO455826669 Apollo Scientific Ltd

3-Chloro-5-(piperidin-1-ylcarbonyl)benzeneboronic acid

  • MDL number MFCD09800859
  • Cas Number : 957120-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-145
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40484

Safety Information

signalword Warning
pcodes P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40484
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart