APO455826672 Apollo Scientific Ltd

8-Bromo-6-nitroimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD09800973
  • Cas Number : 957120-43-3
  • Molecular Weight : 278.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-bromo-6-nitroimidazo12-apyridine-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-bromo-6-nitroimidazo12-apyridine-hydrochloride
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart