APO455826675 Apollo Scientific Ltd

5-(2,3-Dichlorophenyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD07376179
  • Cas Number : 951885-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Carboxy-5-(2,3-dichlorophenyl)-1,3-oxazole, 1-(4-Carboxy-1,3-oxazol-5-yl)-2,3-dichlorobenzene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40547

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40547
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart