APO455826683 Apollo Scientific Ltd

5-Bromo-2-(pyrrolidin-1-ylcarbonyl)pyridine

  • MDL number MFCD09801066
  • Cas Number : 742085-70-7
  • Molecular Weight : 255.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromopyridin-2-yl)(pyrrolidin-1-yl)methanone
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-pyrrolidin-1-ylcarbonylpyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-pyrrolidin-1-ylcarbonylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart