APO455826684 Apollo Scientific Ltd

5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid

  • MDL number MFCD09800892
  • Cas Number : 957062-53-2
  • Molecular Weight : 224.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 3-borono-4-methoxybenzoate, 2-Borono-4-(ethoxycarbonyl)anisole
Purity 95
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-ethoxycarbonyl-2-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/5-ethoxycarbonyl-2-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart