APO455826686 Apollo Scientific Ltd

2-Methoxy-5-(methoxycarbonyl)benzeneboronic acid

  • MDL number MFCD09800877
  • Cas Number : 221006-63-9
  • Molecular Weight : 209.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-borono-4-methoxybenzoate
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-5-methoxycarbonylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-5-methoxycarbonylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart