APO455826688 Apollo Scientific Ltd

5-(Ethoxycarbonyl)thiophene-3-boronic acid

  • MDL number MFCD09800891
  • Cas Number : 957121-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 4-boronothiophene-2-carboxylate, 4-Borono-2-(ethoxycarbonyl)thiophene
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40655

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40655
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart