APO455826692 Apollo Scientific Ltd

2-Bromo-6-nitroaniline

  • MDL number MFCD09263476
  • Cas Number : 59255-95-7
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-74
Synonym 2-Amino-3-bromonitrobenzene
Purity 99
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart