APO455826693 Apollo Scientific Ltd

3-Bromobenzene-1,2-diamine

  • MDL number MFCD09842434
  • Cas Number : 1575-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Synonym 1-Bromo-2,3-diaminobenzene
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromobenzene-12-diamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromobenzene-12-diamine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart