APO455826695 Apollo Scientific Ltd

1,3-Dimethylpyrrolidin-2-one

  • MDL number MFCD01687329
  • Cas Number : 19597-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,3-Dimethyl-2-oxopyrrolidine
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/13-dimethylpyrrolidin-2-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-dimethylpyrrolidin-2-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart