APO455826697 Apollo Scientific Ltd

5-Methylpyridine-2-carbonitrile

  • MDL number MFCD06200830
  • Cas Number : 1620-77-5
  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-77
Synonym 2-Cyano-5-methylpyridine, 5-Methylpicolinonitrile
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 140/2mm
Product Link https://store.apolloscientific.co.uk/product/5-methylpyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylpyridine-2-carbonitrile
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart