APO455826703 Apollo Scientific Ltd

2-Bromo-5-methoxybenzeneboronic acid

  • MDL number MFCD06659859
  • Cas Number : 89694-44-0
  • Molecular Weight : 230.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-216
Synonym 3-Borono-4-bromoanisole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart