APO455826712 Apollo Scientific Ltd

5-Bromo-2-(piperidin-1-ylcarbonyl)pyridine

  • MDL number MFCD08458472
  • Cas Number : 934000-33-6
  • Molecular Weight : 269.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromopicolinoyl)piperidine, (5-Bromopyridin-2-yl)(piperidin-1-yl)methanone
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-piperidin-1-ylcarbonylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-piperidin-1-ylcarbonylpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart