APO455826720 Apollo Scientific Ltd

2-Chloro-5-nitronicotinic acid

  • MDL number MFCD06658402
  • Cas Number : 42959-38-6
  • Molecular Weight : 202.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 140-143
Iupac Name 2-chloro-5-nitronicotinic acid
Inchi Code 1S/C6H3ClN2O4/c7-5-4(6(10)11)1-3(2-8-5)9(12)13/h1-2H,(H,10,11)
Inchi Key WOFZRBCITDVRON-UHFFFAOYSA-N

Description

Synonym 2-Chloro-5-nitropyridine-3-carboxylic acid, 3-Carboxy-2-chloro-5-nitropyridine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 140-143
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-nitronicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-nitronicotinic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart