APO455826723 Apollo Scientific Ltd

Methyl imidazo[1,2-a]pyridine-6-carboxylate hydrochloride

  • MDL number MFCD09801003
  • Cas Number : 957120-91-1
  • Molecular Weight : 212.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-(Methoxycarbonyl)imidazo[1,2-a]pyridine hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-imidazo12-apyridine-6-carboxylate-hydrochloride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-imidazo12-apyridine-6-carboxylate-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart