APO455826726 Apollo Scientific Ltd

2-Amino-5-bromo-4-methoxynicotinonitrile

  • MDL number MFCD09801018
  • Cas Number : 951884-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-5-bromo-4-methoxypyridine-3-carbonitrile, 2-Amino-5-bromo-3-cyano-4-methoxypyridine
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-methoxynicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-5-bromo-4-methoxynicotinonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart